siRNA Details
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053304.1 | NP_445756.1 | 49 | 3904 | ||
GenBank mRNA | BC133728.1 | - | 49 | 3888 | 7125649 | |
KJ696743.1 | - | 5 | 500 | |||
Z78279.1 | - | 49 | 3793 |

More Information
Other Species Targeted

Gene Ontology Categories:
