siRNA Details
Gene Symbol: Hmgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017187.2 | NP_058883.1 | 1 | 27 | ||
GenBank mRNA | BC089854.1 | - | 1 | 27 | 7380208 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008252, BY009449, AK012568.
Gene Ontology Categories:
