siRNA Details
Gene Symbol: Tas1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053305.1 | NP_445757.1 | 4 | 1367 | ||
XM_008764341.1 | XP_008762563.1 | 4 | 1227 | |||
GenBank mRNA | AF127389.1 | - | 4 | 1367 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
