siRNA Details
Gene Symbol: Aadat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017193.1 | NP_058889.1 | 5 | 691 | ||
XM_006253067.2 | XP_006253129.1 | 5 | 779 | |||
GenBank mRNA | BC078864.1 | - | 5 | 687 | 7109210 | |
Z50144.1 | - | 5 | 691 |
