siRNA Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019209.1 | NP_062082.1 | 8, 9 | 921 | ||
GenBank mRNA | BC158690.1 | - | 8, 9 | 921 | 8376316 | |
FQ216026.1 | - | 8, 9 | 916 | |||
FQ216320.1 | - | 8, 9 | 897 | |||
FQ218910.1 | - | 8, 9 | 912 | |||
FQ230458.1 | - | 9, 10 | 766 | |||
FQ233169.1 | - | 8, 9 | 898 | |||
FQ234620.1 | - | 8, 9 | 919 | |||
FQ234801.1 | - | 8, 9 | 897 |
