siRNA Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019209.1 | NP_062082.1 | 3 | 239 | ||
GenBank mRNA | BC158690.1 | - | 3 | 239 | 8376316 | |
DY314066.1 | - | 3 | 239 | |||
EV767503.1 | - | 2 | 197 | |||
FQ216026.1 | - | 3 | 232 | |||
FQ216320.1 | - | 3 | 215 | |||
FQ218910.1 | - | 3 | 230 | |||
FQ230458.1 | - | 3 | 212 | |||
FQ233169.1 | - | 3 | 216 | |||
FQ234620.1 | - | 3 | 237 | |||
FQ234801.1 | - | 3 | 216 | |||
FQ235247.1 | - | 3 | 216 |
