siRNA Details
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017194.1 | NP_058890.1 | 7 | 816 | ||
XM_006246678.2 | XP_006246740.1 | 8 | 934 | |||
XM_008767807.1 | XP_008766029.1 | 8 | 917 | |||
XM_008767808.1 | XP_008766030.1 | 9 | 1583 | |||
GenBank mRNA | X13252.1 | - | 7 | 816 | ||
X74836.1 | - | 7 | 816 |
