siRNA Details
Gene Symbol: Gap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017195.3 | NP_058891.1 | 2 | 295 | ||
GenBank mRNA | BC127502.1 | - | 2 | 143 | 8364847 | |
FQ214107.1 | - | 2 | 130 | |||
M16228.1 | - | 2 | 148 | |||
M16736.1 | - | 2 | 131 | |||
X06338.1 | - | 2 | 295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
