siRNA Details
Gene Symbol: Htt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024357.3 | NP_077333.2 | 33 | 4412 | ||
XM_006251230.2 | XP_006251292.1 | 32 | 4350 | |||
XM_006251232.2 | XP_006251294.1 | 33 | 4398 | |||
XM_008770274.1 | XP_008768496.1 | 31 | 3864 | |||
GenBank mRNA | U18650.1 | - | 33 | 4257 |

More Information
Other Species Targeted

Gene Ontology Categories:
