siRNA Details
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083586.2 | NP_001077055.2 | 1 | 250 | ||
NM_017197.3 | NP_058893.2 | 1 | 250 | |||
XM_008771861.1 | XP_008770083.1 | 1 | 129 | |||
GenBank mRNA | AF090695.1 | - | 1 | 84 | ||
AJ010386.1 | - | 1 | 298 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010160, NM_001110228, NM_001160293, CF536042, AF090697, and others.
Gene Ontology Categories:
