siRNA Details
Gene Symbol: Pak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053306.3 | NP_445758.2 | 1504 | |||
GenBank mRNA | S80221.1 | - | 1423 | |||
U19967.1 | - | 1504 | ||||
U35345.1 | - | 1423 |
Gene Symbol: LOC100910732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751066.2 | XP_003751114.1 | 14 | 1652 | ||
XM_006248473.1 | XP_006248535.1 | 15 | 1781 | |||
XM_008768778.1 | XP_008767000.1 | 14 | 3213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
