siRNA Details
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177321.1 | NP_001170792.1 | 4, 5 | 484 | ||
NM_017200.1 | NP_058896.1 | 3, 4 | 412 | |||
XM_006234440.2 | XP_006234502.1 | 4, 5 | 707 | |||
XM_006234441.2 | XP_006234503.1 | 4, 5 | 1728 | |||
XM_006234444.2 | XP_006234506.1 | 4, 5 | 1741 | |||
XM_008761980.1 | XP_008760202.1 | 4, 5 | 714 | |||
XM_008761981.1 | XP_008760203.1 | 4, 5 | 478 | |||
XM_008761982.1 | XP_008760204.1 | 4, 5 | 708 | |||
XM_008761983.1 | XP_008760205.1 | 4, 5 | 1731 | |||
XM_008761984.1 | XP_008760206.1 | 4, 5 | 1744 | |||
XM_008761985.1 | XP_008760207.1 | 4, 5 | 720 | |||
XM_008761986.1 | XP_008760208.1 | 4, 5 | 714 | |||
GenBank mRNA | BC087020.1 | - | 4, 5 | 484 | 7192382 | |
D10926.1 | - | 3, 4 | 412 | |||
FQ219876.1 | - | 1, 2 | 118 | |||
FQ220339.1 | - | 1, 2 | 112 | |||
FQ220655.1 | - | 1, 2 | 209 | |||
FQ220711.1 | - | 1, 2 | 209 | |||
FQ229364.1 | - | 1, 2 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
