siRNA Details
Gene Symbol: Ahcy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017201.1 | NP_058897.1 | 7, 8 | 875 | ||
GenBank mRNA | M15185.1 | - | 7, 8 | 875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
