siRNA Details
Gene Symbol: Ghrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031577.1 | NP_113765.1 | 4 | 392 | ||
XM_008762304.1 | XP_008760526.1 | 4 | 294 | |||
GenBank mRNA | M73486.1 | - | 4 | 392 | ||
U10156.1 | - | 4 | 294 | |||
U41183.1 | - | 3 | 215 | |||
Z34092.1 | - | 4 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
