siRNA Details
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053307.2 | NP_445759.1 | 2 | 176 | ||
XM_008770745.1 | XP_008768967.1 | 2 | 547 | |||
GenBank mRNA | AY005464.1 | - | 2 | 166 | ||
BC087009.1 | - | 2 | 176 | 7107102 | ||
DQ989019.1 | - | 3 | 208 | |||
DQ989020.1 | - | 3 | 211 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
