siRNA Details
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023090.1 | NP_075578.1 | 12 | 2007 | ||
GenBank mRNA | AJ277828.1 | - | 12 | 2007 |

More Information
Other Species Targeted

Gene Ontology Categories:
