siRNA Details
Gene Symbol: Map6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017204.1 | NP_058900.1 | 4 | 2298 | ||
XM_006229755.2 | XP_006229817.1 | 3 | 2628 | |||
GenBank mRNA | AJ002556.1 | - | 3 | 2317 | ||
BC078848.1 | - | 3 | 2176 | |||
X93495.1 | - | 4 | 2298 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010837, NM_001048167, AK087413, BC053039, AK018193.
Gene Ontology Categories:
