siRNA Details
Gene Symbol: Neurod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019218.2 | NP_062091.1 | 2 | 1092 | ||
GenBank mRNA | AF107728.1 | - | 1 | 1005 | ||
BC092367.1 | - | 2 | 1092 | 7312547 | ||
BC094526.1 | - | 1 | 1309 | 7318277 | ||
D82074.1 | - | 2 | 1102 | |||
D82075.1 | - | 1 | 2569 | |||
D82945.1 | - | 1 | 1005 |
