siRNA Details
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289941.1 | NP_001276870.1 | 7 | 1422 | ||
NM_001289942.1 | NP_001276871.1 | 8 | 1549 | |||
NM_001289943.1 | NP_001276872.1 | 6 | 1213 | |||
NM_019225.2 | NP_062098.1 | 8 | 1539 | |||
GenBank mRNA | AF265360.1 | - | 7 | 1261 | ||
S59158.1 | - | 8 | 1481 | |||
S75687.1 | - | 4 | 765 |

More Information
Other Species Targeted

Gene Ontology Categories:
