siRNA Details
Gene Symbol: Smc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031583.2 | NP_113771.2 | 16 | 1731 | ||
GenBank mRNA | CA511001.1 | - | 1 | 49 | ||
EV776219.1 | - | 4 | 483 | |||
U82626.1 | - | 16 | 1698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
