siRNA Details
Gene Symbol: Notch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024358.1 | NP_077334.1 | 26 | 5238 | ||
GenBank mRNA | M93661.1 | - | 26 | 5238 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Notch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024358.1 | NP_077334.1 | 26 | 5238 | ||
GenBank mRNA | M93661.1 | - | 26 | 5238 |