siRNA Details
Gene Symbol: Notch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024358.1 | NP_077334.1 | 22 | 4058 | ||
GenBank mRNA | M93661.1 | - | 22 | 4058 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010928, AK155137, X68279, D32210, AK155666.
Gene Ontology Categories:
