siRNA Details
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019232.3 | NP_062105.2 | 1, 2 | 137 | ||
GenBank mRNA | BC078843.1 | - | 1, 2 | 91 | 7130800 | |
FM068981.1 | - | 1, 2 | 143 | |||
FM123924.1 | - | 1, 2 | 130 | |||
L01624.1 | - | 1, 2 | 142 |
