siRNA Details
Gene Symbol: Lgals7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022582.2 | NP_072104.2 | 2 | 183 | ||
XM_006228668.2 | XP_006228730.1 | 5 | 441 | |||
XM_006228670.2 | XP_006228732.1 | 4 | 613 | |||
GenBank mRNA | AF036941.1 | - | 2 | 192 | ||
U67883.1 | - | 3 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
