siRNA Details
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017231.1 | NP_058927.1 | 4, 6 | 484 | ||
GenBank mRNA | BC070945.1 | - | 4, 6 | 512 | 7103390 | |
M25758.1 | - | 4, 6 | 484 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK165659, BC056171, AK149745, AK168720, AK187087, and others.
Gene Ontology Categories:
