siRNA Details
Gene Symbol: Vamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057097.2 | NP_476438.1 | 3 | 177 | ||
GenBank mRNA | BC088119.1 | - | 3 | 177 | 7366421 | |
S63830.1 | - | 229 |

More Information
Other Species Targeted

Gene Ontology Categories:
