siRNA Details
Gene Symbol: Hpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017233.1 | NP_058929.1 | 8 | 504 | ||
GenBank mRNA | AF082834.1 | - | 8 | 504 | ||
BC081819.1 | - | 8 | 502 | 7107086 | ||
FQ209393.1 | - | 7 | 658 | |||
FQ209489.1 | - | 7 | 464 | |||
FQ210905.1 | - | 8 | 512 | |||
FQ218378.1 | - | 8 | 512 | |||
FQ218745.1 | - | 8 | 511 | |||
FQ219696.1 | - | 8 | 512 | |||
M18405.1 | - | 7 | 460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
