siRNA Details
Gene Symbol: Ighmbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031586.1 | NP_113774.1 | 3 | 275 | ||
GenBank mRNA | AF199411.1 | - | 3 | 275 | ||
BC099790.1 | - | 3 | 471 | 7934952 |

More Information
Other Species Targeted

Gene Ontology Categories:
