siRNA Details
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053309.1 | NP_445761.1 | 8 | 1135 | ||
GenBank mRNA | AB007689.1 | - | 8 | 1135 | ||
AB007690.1 | - | 8 | 916 |

More Information
Other Species Targeted

Gene Ontology Categories:
