siRNA Details
Gene Symbol: Homer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053310.1 | NP_445762.1 | 3 | 262 | ||
XM_006252890.2 | XP_006252952.1 | 4 | 388 | |||
XM_006252891.2 | XP_006252953.1 | 4 | 385 | |||
XM_008771094.1 | XP_008769316.1 | 4 | 386 | |||
XM_008771095.1 | XP_008769317.1 | 4 | 386 | |||
GenBank mRNA | AB020879.2 | - | 3 | 262 | ||
BC070922.1 | - | 4 | 371 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001146153, NM_011984, BC095961, BC005773, BU054773, and others.
Gene Ontology Categories:
