siRNA Details
Gene Symbol: Vipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017238.1 | NP_058934.1 | 11 | 1185 | ||
GenBank mRNA | U09631.1 | - | 11 | 1185 | ||
Z25885.1 | - | 11 | 1138 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
