siRNA Details
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033987.1 | NP_001029159.1 | 4 | 872 | ||
NM_017246.2 | NP_058942.1 | 4 | 872 | |||
XM_006243229.2 | XP_006243291.1 | 4 | 931 | |||
XM_008766221.1 | XP_008764443.1 | 2 | 70 | |||
GenBank mRNA | BC078860.1 | - | 4 | 872 | 7122708 | |
U37462.1 | - | 4 | 360 | |||
U37463.1 | - | 2 | 81 | |||
U37464.1 | - | 4 | 360 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011840, AK020716, BC028260, AK166137, BC013697, and others.
Gene Ontology Categories:
