siRNA Details
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033987.1 | NP_001029159.1 | 3 | 819 | ||
NM_017246.2 | NP_058942.1 | 3 | 819 | |||
XM_006243229.2 | XP_006243291.1 | 3 | 878 | |||
GenBank mRNA | BC078860.1 | - | 3 | 819 | 7122708 | |
U37462.1 | - | 3 | 307 | |||
U37464.1 | - | 3 | 307 |
