siRNA Details
Gene Symbol: Pou3f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017252.2 | NP_058948.1 | 1 | 1004 | ||
GenBank mRNA | M84645.1 | - | 1 | 1004 | ||
Z11834.1 | - | 1 | 976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
