siRNA Details
Gene Symbol: Ckmt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012738.1 | NP_001012756.1 | 4 | 864 | ||
XM_006234830.2 | XP_006234892.1 | 5 | 820 | |||
GenBank mRNA | BC091335.1 | - | 4 | 864 | 7314893 | |
X59737.1 | - | 5 | 774 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
