siRNA Details
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019242.1 | NP_062115.1 | 5 | 593 | ||
XM_008764643.1 | XP_008762865.1 | 4 | 1212 | |||
GenBank mRNA | BC083560.1 | - | 5 | 659 | 7193329 | |
FQ222510.1 | - | 1 | 17 | |||
J04511.1 | - | 5 | 593 |
