siRNA Details
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019246.1 | NP_062119.1 | 4 | 842 | ||
GenBank mRNA | U36580.1 | - | 4 | 842 |

More Information
Other Species Targeted

Gene Ontology Categories:
