siRNA Details
Gene Symbol: Ptprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168632.1 | NP_001162103.1 | 7 | 936 | ||
XM_008767422.1 | XP_008765644.1 | 3 | 349 |

More Information
Other Species Targeted

Gene Ontology Categories:
