siRNA Details
Gene Symbol: Crhbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139183.2 | NP_631922.1 | 4 | 631 | ||
GenBank mRNA | X58023.1 | - | 4 | 630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
