siRNA Details
Gene Symbol: Bet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019251.2 | NP_062124.1 | 4 | 367 | ||
GenBank mRNA | BC062408.1 | - | 4 | 367 | 5621977 | |
FQ223263.1 | - | 4 | 442 | |||
U42755.1 | - | 4 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
