siRNA Details
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017349.2 | NP_059045.2 | 1, 2 | 56 | ||
NM_145717.1 | NP_663769.1 | 2, 3 | 100 | |||
XM_006242911.2 | XP_006242973.1 | 2, 3 | 257 | |||
XM_006242912.2 | XP_006242974.1 | 2, 3 | 205 | |||
GenBank mRNA | AF129400.1 | - | 2, 3 | 100 | ||
AF233060.1 | - | 1, 2 | 56 | |||
X70062.1 | - | 2, 3 | 64 |
