siRNA Details
Gene Symbol: Dpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019252.1 | NP_062125.1 | 3 | 702 | ||
GenBank mRNA | AB013359.1 | - | 3 | 702 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019252.1 | NP_062125.1 | 3 | 702 | ||
GenBank mRNA | AB013359.1 | - | 3 | 702 |