siRNA Details
Gene Symbol: Psma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017279.1 | NP_058975.1 | 5, 6 | 472 | ||
GenBank mRNA | BC060576.1 | - | 5, 6 | 491 | 6919255 | |
FQ214776.1 | - | 5, 6 | 487 | |||
FQ223872.1 | - | 487 | ||||
J02897.1 | - | 5, 6 | 472 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008944, AK209655, BC026768, AK012119, AK167752, and others.
Gene Ontology Categories:
