siRNA Details
Gene Symbol: Psma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017282.1 | NP_058978.1 | 4 | 331 | ||
GenBank mRNA | BC060575.1 | - | 4 | 352 | 6920479 | |
D10756.1 | - | 4 | 331 | |||
FQ218317.1 | - | 4 | 348 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011967, AK160530, AK160550, BC007181, BC083342, and others.
Gene Ontology Categories:
