siRNA Details
Gene Symbol: Sart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031596.1 | NP_113784.1 | 2 | 403 | ||
GenBank mRNA | AB014722.1 | - | 2 | 403 | ||
BC083551.1 | - | 2 | 376 | 7134496 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
