siRNA Details
Gene Symbol: C1qb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019262.1 | NP_062135.1 | 3 | 620 | ||
GenBank mRNA | FQ227589.1 | - | 3 | 545 | ||
FQ228182.1 | - | 3 | 480 | |||
FQ234810.1 | - | 3 | 559 | |||
X71127.1 | - | 3 | 620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
