siRNA Details
Gene Symbol: Pde1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022710.1 | NP_073201.1 | 9 | 967 | ||
XM_006242386.2 | XP_006242448.1 | 8 | 1539 | |||
GenBank mRNA | AF327906.1 | - | 9 | 960 | ||
FQ212460.1 | - | 9 | 1023 | |||
M94537.1 | - | 9 | 967 |

More Information
Other Species Targeted

Gene Ontology Categories:
