siRNA Details
Gene Symbol: Gabrr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017292.2 | NP_058988.1 | 9 | 1390 | ||
XM_008763610.1 | XP_008761832.1 | 11 | 1376 | |||
XM_008763611.1 | XP_008761833.1 | 8 | 2322 | |||
GenBank mRNA | D38494.1 | - | 9 | 1390 |
