siRNA Details
Gene Symbol: Pcbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007601.2 | NP_001007602.1 | 3 | 237 | ||
XM_006256414.1 | XP_006256476.1 | 4 | 495 | |||
GenBank mRNA | DV724713.1 | - | 1 | 46 | ||
FM060466.1 | - | 3 | 237 | |||
FM067129.1 | - | 3 | 214 | |||
M83740.1 | - | 3 | 433 |

More Information
Other Species Targeted

Gene Ontology Categories:
