siRNA Details
Gene Symbol: Eif2ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031599.2 | NP_113787.1 | 8, 9 | 1614 | ||
XM_006236623.1 | XP_006236685.1 | 8, 9 | 1132 | |||
XM_006236624.2 | XP_006236686.1 | 8, 9 | 1652 | |||
XM_008762977.1 | XP_008761199.1 | 7, 8 | 1246 | |||
GenBank mRNA | AF096835.1 | - | 8, 9 | 1615 |
